Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EMG1 Monoclonal Antibody (OTI1B8)
GREENER_CHOICE

Catalog No. PIMA525111
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525111 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525111 Supplier Invitrogen™ Supplier No. MA525111
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

EMG1 Monoclonal Antibody for Western Blot, ICC/IF, Flow

EMG1 involved in 40S ribosomal subunit biogenesis. It seems to play a role a methylation reaction in pre-rRNA processing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EMG1
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI1B8
Concentration 0.97 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene EMG1
Gene Accession No. Q92979
Gene Alias 18S rRNA (pseudouridine(1248)-N1)-methyltransferase; 18S rRNA (pseudouridine-N1-)-methyltransferase NEP1; 18S rRNA Psi1248 methyltransferase; C2f; EGK_03301; Emg1; EMG1 N1-specific pseudouridine methyltransferase; EMG1 nucleolar protein homolog; EMG1 nucleolar protein homolog (S. cerevisiae); EMG1, N1-specific pseudouridine methyltransferase; essential for mitotic growth 1; gene rich cluster, C2f; Grcc2f; LOC553478 protein; MGC138092 protein; NEP1; nucleolar essential protein 1; nucleolar protein EMG1 homolog; Nucleolar protein EMG1-like protein; probable ribosome biogenesis protein NEP1; protein C2f; Ribosomal RNA small subunit methyltransferase NEP1; ribosome biogenesis protein NEP1; Unknown (protein for MGC:136360); zgc:136360
Gene Symbols EMG1
Host Species Mouse
Immunogen Full length human recombinant protein of EMG1 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10436, 477708
Target Species Human, Canine, Monkey
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.