Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EMG1 Antibody (OTI1B8), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271601H
Description
EMG1 Monoclonal specifically detects EMG1 in Human, Mouse, Rat, Canine, Monkey, Primate samples. It is validated for Western Blot.Specifications
| EMG1 | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| EMG1 | |
| Full length recombinant protein of human EMG1 (NP_006322) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey, Primate | |
| IgG2b |
| Western Blot | |
| OTI1B8 | |
| Western Blot | |
| 18S rRNA (pseudouridine-N1-)-methyltransferase NEP1, BWCNS, C2F18S rRNA Psi1248 methyltransferase, EMG1 nucleolar protein homolog (S. cerevisiae), essential for mitotic growth 1, FLJ60792, Grcc2f, NEP1EC 2.1.1.-, Nucleolar protein EMG1 homolog, probable ribosome biogenesis protein NEP1, Protein C2f, Ribosome biogenesis protein NEP1 | |
| Mouse | |
| Affinity Purified | |
| DNA replication Transcription Translation and Splicing | |
| 10436.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction