Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EMG1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565231
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565231 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565231 Supplier Invitrogen™ Supplier No. PA565231
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PSDGFKPRERS GGEQAQDWDA LPPKRPRLGA GNKIGGRRLI VVLEGASLET VKVGKTYELL NCDKHKSILL KNGRDP Highest antigen sequence identity to the following orthologs - mouse 78%, rat 83%.

EMG1 involved in 40S ribosomal subunit biogenesis. It seems to play a role a methylation reaction in pre-rRNA processing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EMG1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene EMG1
Gene Accession No. Q92979
Gene Alias 18S rRNA (pseudouridine(1248)-N1)-methyltransferase; 18S rRNA (pseudouridine-N1-)-methyltransferase NEP1; 18S rRNA Psi1248 methyltransferase; C2f; EGK_03301; Emg1; EMG1 N1-specific pseudouridine methyltransferase; EMG1 nucleolar protein homolog; EMG1 nucleolar protein homolog (S. cerevisiae); EMG1, N1-specific pseudouridine methyltransferase; essential for mitotic growth 1; gene rich cluster, C2f; Grcc2f; LOC553478 protein; MGC138092 protein; NEP1; nucleolar essential protein 1; nucleolar protein EMG1 homolog; Nucleolar protein EMG1-like protein; probable ribosome biogenesis protein NEP1; protein C2f; Ribosomal RNA small subunit methyltransferase NEP1; ribosome biogenesis protein NEP1; Unknown (protein for MGC:136360); zgc:136360
Gene Symbols EMG1
Host Species Rabbit
Immunogen Recombinant Human EMG1. Recombinant protein control fragment (Product #RP-104702).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10436
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.