Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EMID1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543542
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543542 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543542 Supplier Invitrogen™ Supplier No. PA543542
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: TMIGLYEPEL GSGAGPAGTG TPSLLRGKRG GHATNYRIVA PRSRDERG Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 92%; Horse: 100%; Human: 100%; Mouse: 100%; Rat: 100%.

EMID1 contains 1 collagen-like domain and 1 EMI domain. The exact function of EMID1 remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EMID1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene EMID1
Gene Accession No. Q96A84
Gene Alias AW122071; CO-5; EMI domain containing 1; EMI domain-containing protein 1; EMI5; Emid1; Emilin and multimerin domain-containing protein 1; Emu1; RGD1565846
Gene Symbols EMID1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human EMID1 (aa 394-441).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 129080
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.