Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EMID1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983302
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3302 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3302 Supplier Bioss Supplier No. BS14581R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EMID1 contains 1 collagen-like domain and 1 EMI domain. The exact function of EMID1 remains unknown.

Specifications

Antigen EMID1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene EMID1
Gene Accession No. Q96A84
Gene Alias AW122071; BOS_16714; CO-5; EMI domain containing 1; EMI domain-containing protein 1; EMI5; Emid1; Emilin and multimerin domain-containing protein 1; Emu1; RGD1565846
Gene Symbols EMID1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human EMID1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 129080
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.