Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EMID2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP259788 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP259788 100 μL
NB393135 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP259788 Supplier Novus Biologicals Supplier No. NBP259788
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EMID2 Polyclonal specifically detects EMID2 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen EMID2
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200
Formulation PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
Gene Alias COL26A1, EMI domain containing 2, EMI domain-containing protein 2, Emilin and multimerin domain-containing protein 2, emilin and multimerin-domain containing protein 2, EMU2, Emu2type XXVI, alpha 1, hEmu2, KIAA1299, MGC129848, putative emu2, SH2B, SH2B1
Gene Symbols EMID2
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to the amino acid sequence:ANLVSYRTLIRPTYRVSYRTVTVLEWRCCPGFTGSNCDEECMNCTRLSDMSERLTTLEAKVLLLEAAERPSSPDNDLPAPESTPPTWNEDFLPDAIPLAHPVP
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 136227
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.