Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EMID2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP169681
Description
EMID2 Polyclonal specifically detects EMID2 in Human samples. It is validated for Western Blot.Specifications
EMID2 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
COL26A1, EMI domain containing 2, EMI domain-containing protein 2, Emilin and multimerin domain-containing protein 2, emilin and multimerin-domain containing protein 2, EMU2, Emu2type XXVI, alpha 1, hEmu2, KIAA1299, MGC129848, putative emu2, SH2B, SH2B1 | |
Rabbit | |
45 kDa | |
100 μL | |
Primary | |
Zebrafish: 83%; Guinea pig: 83%. | |
Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
IgG |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
Q96A83 | |
EMID2 | |
Synthetic peptides corresponding to EMID2(EMI domain containing 2) The peptide sequence was selected from the C terminal of EMID2. Peptide sequence LAGERGTVGPSGEPGVKGEEGEKAATAEGEGVQQLREALKILAERVLILE. | |
Affinity purified | |
RUO | |
136227 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction