Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EMILIN1 Antibody (C11A8), mFluor Violet 450 SE, Novus Biologicals™

Rat Monoclonal Antibody
$585.00
Specifications
Antigen | EMILIN1 |
---|---|
Clone | C11A8 |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Frozen) |
Classification | Monoclonal |
Conjugate | mFluor Violet 450 SE |
Description
EMILIN1 Monoclonal antibody specifically detects EMILIN1 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Frozen)Specifications
EMILIN1 | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Frozen) | |
mFluor Violet 450 SE | |
Rat | |
Mouse | |
11117 | |
IgG2a κ | |
Protein G purified |
C11A8 | |
Monoclonal | |
Purified | |
RUO | |
DKFZp586M121, elastin microfibril interface-located protein 1, elastin microfibril interfacer 1, EMI, EMILIN, EMILIN-1, gp115 | |
recombinant mouse Emilin-1 | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title