Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EMILIN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551745
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551745 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551745 Supplier Invitrogen™ Supplier No. PA551745
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PQSIMYRRFL RPRYRVAYKT VTDMEWRCCQ GYGGDDCAES PAPALGPASS TPRPLARPAR PNLSGSSAGS PLSGLGGEGP GESEKVQQLE EQVQSLTKEL QGLRGVLQGL SGRLAEDVQR AVETAFNGRQ QPADAAARPG VHETLNE Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 74%.

This gene encodes a protein that is one of the two components of elastic fibers. The encoded protein is rich in hydrophobic amino acids such as glycine and proline, which form mobile hydrophobic regions bounded by crosslinks between lysine residues. Deletions and mutations in this gene are associated with supravalvular aortic stenosis (SVAS) and autosomal dominant cutis laxa. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EMILIN1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.20 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene EMILIN1
Gene Accession No. Q9Y6C2
Gene Alias 5830419M17Rik; AW229038; BB105748; elastin microfibril interface located protein 1; elastin microfibril interface-located protein 1; Elastin microfibril interfacer 1; EMI; EMILIN; EMILIN1; EMILIN-1; gp115; LOW QUALITY PROTEIN: EMILIN-1
Gene Symbols EMILIN1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human EMILIN1. Recombinant protein control fragment (Product #RP-101050).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11117
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.