Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EMILIN2 Antibody (828B3B3) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$243.50 - $560.50
Specifications
Antigen | EMILIN2 |
---|---|
Clone | 828B3B3 |
Concentration | LYOPH |
Dilution | Western Blot 2-5ug/ml, Immunohistochemistry-Frozen |
Applications | Western Blot, Immunohistochemistry (Frozen) |
Description
EMILIN2 Monoclonal specifically detects EMILIN2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
EMILIN2 | |
LYOPH | |
Western Blot, Immunohistochemistry (Frozen) | |
Unconjugated | |
Mouse | |
Human | |
84034 | |
Recombinant gC1q domain of EMILIN2 | |
Primary | |
Centrifuge vial prior to opening. Reconstitute in sterile water to a concentration of 0.1-1.0 mg/ml. |
828B3B3 | |
Western Blot 2-5ug/ml, Immunohistochemistry-Frozen | |
Monoclonal | |
Purified | |
RUO | |
Elastin microfibril interface-located protein 2, elastin microfibril interfacer 2EMILIN-2, extracellular glycoprotein EMILIN-2, FLJ33200, FOAP-10, Protein FOAP-10 | |
EMILIN2 | |
IgG2 κ | |
Protein G purified | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title