Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ EMILIN2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA584657
Description
Immunogen sequence: RMLNGRLDNE FDRLIVPEPD VDFDAKWNEL DARINVTEKN AEEHCFYIEE TLRGAINGEV GDLKQLVDQK IQSLEDRLGS VLLQMTNNT.
EMILIN2 is a protein coding gene. May be responsible for anchoring smooth muscle cells to elastic fibers, and may be involved not only in the formation of the elastic fiber, but also in the processes that regulate vessel assembly. Has cell adhesive capacity.
Specifications
EMILIN2 | |
Polyclonal | |
Unconjugated | |
EMILIN2 | |
elastin microfibril interface-located protein 2; Elastin microfibril interfacer 2; EMILIN2; EMILIN-2; extracellular glycoprotein EMILIN-2; FLJ33200; FOAP-10; Protein FOAP-10 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84034 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BXX0 | |
EMILIN2 | |
Recombinant protein corresponding to Human EMILIN2. Recombinant protein control fragment (Product #RP-108499). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction