Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ EML 425
GREENER_CHOICESDP

Catalog No. 564650 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
56-465-0 50 mg
56-461-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 56-465-0 Supplier Tocris Bioscience™ Supplier No. 5646/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Reversible and non-competitive CBP/p300 inhibitor

EML 425 is a reversible and non-competitive CBP/p300 inhibitor (IC50 values are 1.1 and 2.9 μM, respectively). Cell permeable.

Specifications

Chemical Name or Material 5-[(4-Hydroxy-2,6-dimethylphenyl)methylene]-1,3-bis(phenylmethyl)-2,4,6(1H,3H,5H)-pyrimidinetrione
CAS 1675821-32-5
Quantity 50 mg
Target Histone Acetyltransferase Inhibitors
Molecular Formula C27H24N2O4
Formula Weight 440.49
Purity >98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.