Learn More
Invitrogen™ EML1 Polyclonal Antibody

Description
Recommended positive controls: A549, HeLa, HepG2, mouse brain. Predicted reactivity: Mouse (93%), Rat (93%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | EML1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.62 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 |
| Gene | EML1 |
| Gene Accession No. | O00423, Q05BC3 |
| Gene Alias | 1110008N23Rik; A930030P13Rik; AA171013; AI847476; AI853955; echinoderm microtubule associated protein like 1; echinoderm microtubule associated protein-like protein 1; echinoderm microtubule-associated protein-like 1; ELP79; EMAP; EMAP1; EMAP-1; EMAPL; EMAPL1; Eml1; HuEMAP; huEMAP-1 |
| Gene Symbols | EML1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.