Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EML2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530493
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530493 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530493 Supplier Invitrogen™ Supplier No. PA530493
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: A549. Predicted reactivity: Mouse (94%), Rat (93%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

May modify the assembly dynamics of microtubules, such that microtubules are slightly longer, but more dynamic. Target protein is a microtubule-associated protein that binds to the delta2 glutamate receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EML2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.97 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene EML2
Gene Accession No. O95834
Gene Alias 1600029N02Rik; echinoderm microtubule associated protein like 2; echinoderm microtubule-associated protein-like 2; echinoderm MT-associated protein (EMAP)-like protein 70; ELP70; EMAP2; EMAP-2; EMAPL2; EML2; microtubule-associated protein like echinoderm EMAP
Gene Symbols EML2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human EML2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24139
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.