Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EML2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA592258
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA592258 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA592258 Supplier Invitrogen™ Supplier No. PA592258
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LHVLGLGVFD RAVCCVGFSK SNGGNLLCAV DESNDHMLSV WDWAKETKVV DVKCSNEAVL VATFHPTDPT VLITCGKSHI YFWTLEGGSL SKRQGLFEKH EKPKYVLCVT FLEGGDVVTG DSGGNLYVWG KGGNRITQAV LGAHDGGVFG LCALRDGTLV SGGGRDRRVV LWGSDYSKLQ EVEVPEDFGP VRTVAEGHGD TLYVGTTRNS ILQGSVHTGF SLLVQGHVEE LWGLATHPSR AQFVTCGQDK LVHLWSSDSH QPLWSR; Positive Samples: Mouse heart, Mouse kidney; Cellular Location: Cytoplasm, cytoskeleton, spindle.

May modify the assembly dynamics of microtubules, such that microtubules are slightly longer, but more dynamic. Target protein is a microtubule-associated protein that binds to the delta2 glutamate receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EML2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.44 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene EML2
Gene Accession No. O95834, Q7TNG5
Gene Alias 1600029N02Rik; echinoderm microtubule associated protein like 2; echinoderm microtubule-associated protein-like 2; echinoderm MT-associated protein (EMAP)-like protein 70; ELP70; EMAP2; EMAP-2; EMAPL2; EML2; microtubule-associated protein like echinoderm EMAP
Gene Symbols EML2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 150-415 of human EML2 (NP_0362871).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24139, 72205
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.