Learn More
Invitrogen™ EML2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5116573
Description
Positive Samples: Mouse brain, Mouse liver Immunogen sequence: LHVLGLGVFD RAVCCVGFSK SNGGNLLCAV DESNDHMLSV WDWAKETKVV DVKCSNEAVL VATFHPTDPT VLITCGKSHI YFWTLEGGSL SKRQGLFEKH EKPKYVLCVT FLEGGDVVTG DSGGNLYVWG KGGNRITQAV LGAHDGGVFG LCALRDGTLV SGGGRDRRVV LWGSDYSKLQ EVEVPEDFGP VRTVAEGHGD TLYVGTTRNS ILQGSVHTGF SLLVQGHVEE LWGLATHPSR AQFVTCGQDK LVHLWSSDSH QPLWSR.
May modify the assembly dynamics of microtubules, such that microtubules are slightly longer, but more dynamic. Target protein is a microtubule-associated protein that binds to the delta2 glutamate receptor.
Specifications
| EML2 | |
| Polyclonal | |
| Unconjugated | |
| EML2 | |
| 1600029N02Rik; echinoderm microtubule associated protein like 2; echinoderm microtubule-associated protein-like 2; echinoderm MT-associated protein (EMAP)-like protein 70; ELP70; EMAP2; EMAP-2; EMAPL2; EML2; microtubule-associated protein like echinoderm EMAP | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 72205 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.84 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q7TNG5 | |
| EML2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 150-415 of human EML2 (NP_0362871). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.