Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EML3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116778
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116778 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116778 Supplier Invitrogen™ Supplier No. PA5116778
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: THP-1 Immunogen sequence: HYRGHTDCVR CLAVHPDGVR VASGQTAGVD KDGKPLQPVV HIWDSETLLK LQEIGLGAFE RGVGALAFSA ADQGAFLCVV DDSNEHMLSV WDCSRGMKLA EIKSTNDSVL AVGFNPRDSS CIVTSGKSHV HFWNWSGGVG VPGNGTLTRK QGVFGKYKKP KFIPCFVFLP DGDILTGDSE GNILTWGRSP SDSKTPGRGG AKETYGIVAQ AHAHEGSIFA LCLRRDGTVL S.

EML3 gene ontology annotations related to this gene include microtubule cytoskeleton organization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EML3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.53 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene EML3
Gene Accession No. Q32P44
Gene Alias BC022146; echinoderm microtubule associated protein like 3; echinoderm microtubule-associated protein-like 3; echinoderm microtubule-associated protein-like 3-like; ELP95; EMAP like 3; EMAP-3; Eml3; LOC102551660
Gene Symbols EML3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 300-530 of human EML3 (NP_6949972).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 256364
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.