Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EML3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570124
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570124 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570124 Supplier Invitrogen™ Supplier No. PA570124
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

EML3 gene ontology annotations related to this gene include microtubule cytoskeleton organization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EML3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene EML3
Gene Accession No. Q32P44
Gene Alias BC022146; echinoderm microtubule associated protein like 3; echinoderm microtubule-associated protein-like 3; echinoderm microtubule-associated protein-like 3-like; ELP95; EMAP like 3; EMAP-3; Eml3; LOC102551660
Gene Symbols EML3
Host Species Rabbit
Immunogen synthetic peptide directed towards the N terminal region of human EML3.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 256364
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.