Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EML3 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$148.20 - $378.00
Specifications
Antigen | EML3 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
EML3 Polyclonal antibody specifically detects EML3 in Human samples. It is validated for Western BlotSpecifications
EML3 | |
Polyclonal | |
Purified | |
RUO | |
Human | |
echinoderm microtubule associated protein like 3 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 300-530 of human EML3 (NP_694997.2). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
Rabbit | |
Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers | |
PBS, 50% glycerol, pH7.3 | |
256364 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title