Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EML4 Antibody, Alexa Fluor™ 700, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP105974AF700
Description
EML4 Polyclonal antibody specifically detects EML4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
EML4 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
C2orf2, echinoderm microtubule associated protein like 4, echinoderm microtubule-associated protein-like 4, ELP120, EMAP-4, EMAPL4, FLJ10942, FLJ32318, Restrictedly overexpressed proliferation-associated protein, Ropp 120, ROPP120DKFZp686P18118 | |
The immunogen recognized by this antibody maps to a region between residue 931 and 981of human echinoderm microtubule associated protein like 4 using the numbering given in entry NP_061936.2 (GeneID 27436). | |
0.1 mL | |
Cell Cycle and Replication | |
27436 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction