Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EML4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP238319
Description
EML4 Polyclonal specifically detects EML4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
EML4 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
Q9HC35 | |
EML4 | |
This antibody was developed against a recombinant protein corresponding to amino acids: PQPSSQPLQIHRQTPESKNATPTKSIKRPSPAEKSHNSWENSDDSRNKLSKIPSTPKLIPKVTKTADKHKDVIINQEGEY | |
0.1 mL | |
Cell Cycle and Replication | |
27436 | |
Human | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
C2orf2, echinoderm microtubule associated protein like 4, echinoderm microtubule-associated protein-like 4, ELP120, EMAP-4, EMAPL4, FLJ10942, FLJ32318, Restrictedly overexpressed proliferation-associated protein, Ropp 120, ROPP120DKFZp686P18118 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction