Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EML5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563116
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563116 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563116 Supplier Invitrogen™ Supplier No. PA563116
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: THVTNVRWTY DDSMLVTLGG TDMSLMVWTN EMEGYREKRP CDSEESDI Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 83%.

EML5 may modify the assembly dynamics of microtubules, such that microtubules are slightly longer, but more dynamic.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EML5
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene EML5
Gene Accession No. Q05BV3
Gene Alias BC027154; C130068M19Rik; echinoderm microtubule associated protein like 5; echinoderm microtubule-associated protein-like 5; EMAP like 5; EMAP-2; EMAP-5; EMAP-like protein 5; EML5; LOW QUALITY PROTEIN: echinoderm microtubule-associated protein-like 5
Gene Symbols EML5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human EML5. Recombinant protein control fragment (Product #RP-101434).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 161436
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.