Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EMMPRIN/CD147 Antibody (041), DyLight 350, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290540UV
Description
EMMPRIN/CD147 Monoclonal specifically detects EMMPRIN/CD147 in Mouse samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.Specifications
EMMPRIN/CD147 | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
No cross-reactivity in ELISA with: Human Basigin/ BSG/ CD147 | |
Store at 4°C in the dark. | |
IgG |
ELISA, Immunofluorescence | |
41 | |
ELISA, Immunocytochemistry/Immunofluorescence | |
5F7, basigin, basigin (Ok blood group), CD147, CD147 antigen, Collagenase stimulatory factor, EMMPRINTCSF, Extracellular matrix metalloproteinase inducer, Leukocyte activation antigen M6, M6, OK, OK blood group antigen, Tumor cell-derived collagenase stimulatory factor | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse EMMPRIN/CD147 (NP_001070652.1; Met 1-Arg 209). | |
0.1 mL | |
Cancer, Signal Transduction, Vision | |
682 | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction