Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EMMPRIN/CD147 Antibody (8D6), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP234543APC
Description
EMMPRIN/CD147 Monoclonal specifically detects EMMPRIN/CD147 in Human samples. It is validated for Flow Cytometry.Specifications
EMMPRIN/CD147 | |
Monoclonal | |
APC | |
5F7, basigin, basigin (Ok blood group), CD147, CD147 antigen, Collagenase stimulatory factor, EMMPRINTCSF, Extracellular matrix metalloproteinase inducer, Leukocyte activation antigen M6, M6, OK, OK blood group antigen, Tumor cell-derived collagenase stimulatory factor | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
This monoclonal antibody recognizes extracellular epitope 2 within the N-terminal Ig domain of human CD147. It is expressed more intensely on thymocytes than on mature peripheral blood T cells. CD147 is important in spermatogenesis, embryo implantation, neural network formation, and tumor progression. It stimulates the production of interstitial collagenase, gelatinase A, stromelysin-1 and various metalloproteinases (MMPs) by fibroblasts. These enzymes are important factors in cancer invasion and metastasis. | |
Store at 4C in the dark. | |
IgG1 κ |
Flow Cytometry | |
8D6 | |
Flow Cytometry | |
BSG | |
Human T cell line Molt 13 | |
0.1 mL | |
Cancer, Signal Transduction, Vision | |
682 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction