Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EMMPRIN/CD147 Antibody (BSG/963), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311531AF594
Description
EMMPRIN/CD147 Monoclonal antibody specifically detects EMMPRIN/CD147 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
EMMPRIN/CD147 | |
Monoclonal | |
Alexa Fluor 594 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
BSG/963 | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
5F7, basigin, basigin (Ok blood group), CD147, CD147 antigen, Collagenase stimulatory factor, EMMPRINTCSF, Extracellular matrix metalloproteinase inducer, Leukocyte activation antigen M6, M6, OK, OK blood group antigen, Tumor cell-derived collagenase stimulatory factor | |
Recombinant human EMMPRIN/CD147 protein (Uniprot: P35613) | |
0.1 mL | |
Cancer, Signal Transduction, Vision | |
682 | |
Store at 4°C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction