Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EMMPRIN/CD147 Antibody (OTI9H5), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270278PE
Description
EMMPRIN/CD147 Monoclonal specifically detects EMMPRIN/CD147 in Human, Rat, Canine, Primate, Monkey samples. It is validated for Flow Cytometry.Specifications
EMMPRIN/CD147 | |
Monoclonal | |
PE | |
PBS with 0.05% Sodium Azide | |
BSG | |
Full length recombinant protein of human EMMPRIN/CD147 (NP_001719) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Rat, Canine, Primate, Monkey | |
IgG2b |
Flow Cytometry | |
OTI9H5 | |
Flow Cytometry | |
5F7, basigin, basigin (Ok blood group), CD147, CD147 antigen, Collagenase stimulatory factor, EMMPRINTCSF, Extracellular matrix metalloproteinase inducer, Leukocyte activation antigen M6, M6, OK, OK blood group antigen, Tumor cell-derived collagenase stimulatory factor | |
Mouse | |
Affinity Purified | |
Cancer, Signal Transduction, Vision | |
682.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction