Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EMMPRIN/CD147 Antibody (OX-47) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 6 publications
Supplier: Novus Biologicals NB10064848
Description
EMMPRIN/CD147 Monoclonal specifically detects EMMPRIN/CD147 in Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, CyTOF-ready.Specifications
EMMPRIN/CD147 | |
Monoclonal | |
1.0 mg/mL | |
Western Blot 1:100 - 1:2000, Flow Cytometry 1 ug/mL, Immunohistochemistry 1:100 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Paraffin 1:100 - 1:500, Immunohistochemistry-Frozen 1:100 - 1:500, CyTOF-ready | |
5F7, basigin, basigin (Ok blood group), CD147, CD147 antigen, Collagenase stimulatory factor, EMMPRINTCSF, Extracellular matrix metalloproteinase inducer, Leukocyte activation antigen M6, M6, OK, OK blood group antigen, Tumor cell-derived collagenase stimulatory factor | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
NB100-64848 recognizes a membrane glycoprotein of MW 42-48 kD expressed at high levels on activated lymphocytes but lower levels on resting cells. The antigen is also present on some endothelia. The antigen contains two Ig-like domains and an unusual transmembrane domain with a charged residue. It is abundant on activated lymphocytes: it is the homologue of the chicken blood brain barrier antigen HT7. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
OX-47 | |
Unconjugated | |
P26453 | |
BSG | |
T blasts from a mixed lymphocyte reaction. | |
0.1 mL | |
Cancer, Signal Transduction, Vision | |
682 | |
Rat | |
Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction