Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EMMPRIN/CD147 Rabbit anti-Human, Monkey, Clone: 125, Novus Biologicals™

Rabbit Monoclonal Antibody
$390.00 - $560.50
Specifications
Antigen | EMMPRIN/CD147 |
---|---|
Clone | 125 |
Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:25-1:100, ELISA 1:5000-1:10000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50-1:1000, Immunoprecipitation 4-6 ul/mg of lysate, Immunohistochemistry-Paraffin 1:2500-1:10000, Immunohistochemistry-Frozen 1:500-1:2000 |
Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
Classification | Monoclonal |
Description
EMMPRIN/CD147 Monoclonal specifically detects EMMPRIN/CD147 in Human, Monkey samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
EMMPRIN/CD147 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:25-1:100, ELISA 1:5000-1:10000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50-1:1000, Immunoprecipitation 4-6 ul/mg of lysate, Immunohistochemistry-Paraffin 1:2500-1:10000, Immunohistochemistry-Frozen 1:500-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Human, Monkey | |
5F7, basigin, basigin (Ok blood group), CD147, CD147 antigen, Collagenase stimulatory factor, EMMPRINTCSF, Extracellular matrix metalloproteinase inducer, Leukocyte activation antigen M6, M6, OK, OK blood group antigen, Tumor cell-derived collagenase stimulatory factor | |
Recombinant Human EMMPRIN/CD147 protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
125 | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
Unconjugated | |
Rabbit | |
Cancer, Signal Transduction, Vision | |
0.2 um filtered solution in PBS | |
682 | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title