Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EMP/MAEA Mouse anti-Human, Mouse, Rat, Clone: 730340, R&D Systems™

Catalog No. MAB7288SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB7288SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB7288SP Supplier R&D Systems Supplier No. MAB7288SP
This item is not returnable. View return policy

Mouse Monoclonal Antibody

EMP/MAEA Monoclonal antibody MAB7288SP specifically detects EMP/MAEA in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Western Blotting

Specifications

Antigen EMP/MAEA
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 730340
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/mL, Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q7L5Y9
Gene Alias EMLP, EMPCell proliferation-inducing gene 5 protein, Erythroblast macrophage protein, HLC-10, Human lung cancer oncogene 10 protein, lung cancer-related protein 10, macrophage erythroblast attacher, MAEA, PIG5, proliferation-inducing gene 5
Gene Symbols MAEA
Host Species Mouse
Immunogen E. coli-derived recombinant human EMP/MAEA Met12-Ser66 Accession # Q7L5Y9
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10296
Test Specificity Detects human EMP/MAEA in direct ELISAs and Western blots.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.