Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EMP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566147
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566147 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566147 Supplier Invitrogen™ Supplier No. PA566147
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GVVEKLSVLKR KAVESIQAED ESAKLCKRRI EHLKEHSSDQ PAAASVWKRK RMDRMMVEHL LRCGYYNTAV KL Highest antigen sequence identity to the following orthologs - mouse 99%, rat 99%.

This gene product mediates the attachment of erythroblasts to macrophages. This attachment promotes terminal maturation and enucleation of erythroblasts, presumably by suppressing apoptosis. This protein is an integral membrane protein with the N-terminus on the extracellular side and the C-terminus on the cytoplasmic side of the cell. Two immunologically related isoforms of erythroblast macrophage protein with apparent molecular weights of 33 kD and 36 kD were detected in macrophage membranes, this gene encodes the larger isoform. Multiple alternatively spliced transcript variants encoding distinct isoforms have been found for this gene, but the biological validity of some variants has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EMP
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MAEA
Gene Accession No. Q7L5Y9
Gene Alias 1110030D19Rik; Cell proliferation-inducing gene 5 protein; E3 ubiquitin-protein transferase MAEA; EMLP; EMP; Erythroblast macrophage protein; GID complex subunit 9, FYV10 homolog; GID9; HLC10; HLC-10; Human lung cancer oncogene 10 protein; lung cancer-related protein 10; macrophage erythroblast attacher; MAEA; P44EMLP; PIG5
Gene Symbols MAEA
Host Species Rabbit
Immunogen Recombinant Human EMP. Recombinant protein control fragment (Product #RP-106828).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10296
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.