Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EMP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597945
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597945 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597945 Supplier Invitrogen™ Supplier No. PA597945
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EMP Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

This gene product mediates the attachment of erythroblasts to macrophages. This attachment promotes terminal maturation and enucleation of erythroblasts, presumably by suppressing apoptosis. This protein is an integral membrane protein with the N-terminus on the extracellular side and the C-terminus on the cytoplasmic side of the cell. Two immunologically related isoforms of erythroblast macrophage protein with apparent molecular weights of 33 kD and 36 kD were detected in macrophage membranes, this gene encodes the larger isoform. Multiple alternatively spliced transcript variants encoding distinct isoforms have been found for this gene, but the biological validity of some variants has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EMP
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 6 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene MAEA
Gene Accession No. Q7L5Y9
Gene Alias 1110030D19Rik; Cell proliferation-inducing gene 5 protein; E3 ubiquitin-protein transferase MAEA; EMLP; EMP; Erythroblast macrophage protein; GID complex subunit 9, FYV10 homolog; GID9; HLC10; HLC-10; Human lung cancer oncogene 10 protein; lung cancer-related protein 10; macrophage erythroblast attacher; MAEA; P44EMLP; PIG5
Gene Symbols MAEA
Host Species Rabbit
Immunogen Recombinant Human Macrophage erythroblast attacher protein (299-356AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10296
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.