Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EMP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA548598
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA548598 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA548598 Supplier Invitrogen™ Supplier No. PA548598
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EMP2 Polyclonal Antibody for Western Blot, IHC (P)

Epithelial membrane protein-2 (EMP2) is a member of the four transmembrane superfamily (TM4SF) and is thought to mediate trafficking of diverse proteins such as alpha6beta1 integrin and MHC class I to lipid raft microdomains. EMP2 has also recently been recognized as a putative tumor suppressor gene in certain model systems.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EMP2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene EMP2
Gene Accession No. O88662, P54851
Gene Alias EMP2; EMP-2; Epithelial membrane protein 2; MGC9056; Protein XMP; Unknown (protein for MGC:137044); XMP
Gene Symbols EMP2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 100-129 amino acids from the C-terminal region of human EMP2.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13731, 2013
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.