Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EMP3 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986843
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6843 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6843 Supplier Bioss Supplier No. BS5836R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Probably involved in cell proliferation and cell-cell interactions.

Specifications

Antigen EMP3
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene EMP3
Gene Accession No. P54852
Gene Alias EMP3; EMP-3; Epithelial membrane protein 3; H4; H-4; Hematopoietic neural membrane protein 1; HNMP-1; MI-35; Protein YMP; YMP
Gene Symbols EMP3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human EMP3.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2014
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.