Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ EMPA
GREENER_CHOICESDP

Catalog No. 455810 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-581-0 10 mg
45-585-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 45-581-0 Supplier R&D Systems™ Supplier No. 4558/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Highly potent and selective OX2 antagonist; brain penetrant

Highly potent, selective OX2 receptor antagonist (IC50 values are 2.3 nM and 1900 nM for OX2 and OX1 respectively). Displays negligible or no inhibition of a panel of 80 receptors. Blocks orexin-B- and orexin-A-invoked calcium mobilization in hOX2-expressing CHO cells (IC50 values are 7.9 nM and 8.8 nM respectively); reverses orexin-B-induced hyperlocomotion in mice. Brain penetrant.

Chemical Identifiers

CAS 680590-49-2
Molecular Formula C23H26N4O4S

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 454.54
Purity >98%
Molecular Weight (g/mol) 454.545
Chemical Name or Material EMPA
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.