Learn More
CDS Analytical™ Empore™ C18 StageTips
SDP
C18 bonded silica StageTips are SPE micropipette tips for the desalting of large peptides and proteins, as well as peptide fractionation.
Supplier:  CDS Analytical™ 70201910013EA
 View more versions of this product
Catalog No. 13-110-055
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
The CDS™ Empore™ StageTips C18 bonded silica solid phase extraction micropipette tips have two layers for increased capacity.
  • Combines good retention capacity with high recoveries
  • Recommended for desalting of large peptides and proteins and peptide fractionation at acidic, neutral, and basic pH conditions
Specifications
Specifications
12 μm
C18 (Octadecyl, ODS)
96 Pk.
Empore
200 μL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.