Learn More
Fisherbrand™ Empty Click Lock Container
SDP
Catalog No. 23101011 Shop All Fisherbrand Products
Click to view available options
Capacity (Metric):
20 mL
40 mL
60 mL
90 mL
120 mL
This item is not returnable. View return policy
This item is not returnable. View return policy
Empty Click Lock Container designed for safe, leak-resistant transport and storage of fluids.
  • Designed for safe, leak-resistant transport and storage of fluids
  • Screw-top containers have knurled cap and base for easy handling
  • Containers are graduated and made of durable clarified polypropylene
Specifications
Product Type Specimen Container Vial
Closure Included Yes
Graduated Yes
Material Polypropylene
Shape Cylindrical
Sterility Non-sterile
Autoclavable Not Autoclavable
Capacity (Metric) 60 mL
Color Clear
No. per Case 400
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.