Learn More
Scientific Cell Company Empty Filter Mount
Shop All Scientific Cell Company Inc. Products
Click to view available options
Type:
Empty Filter Mount
Description
- Solid
- Individual filter
- Type: Glass
- Light Path: Nuetral Density
- Packed in high-quality storage case
Specifications
Specifications
| Type | Empty Filter Mount |
| For Use With (Application) | Testing the photometric accuracy (0 Abs) |