Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Empty Tube for Thermal Desorption

Catalog No. 111008795 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Packaging Quantity:
10/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity
11-100-8795 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-8795 Supplier MilliporeSigma™ Supelco™ Supplier No. 21822U
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Features an etched, unique number for sample identification

  • Preconditioned
  • Screens included
  • Packed with high quality adsorbents
  • Sealed in a TDS3 Storage Container
TRUSTED_SUSTAINABILITY

Specifications

Material Stainless Steel
For Use With (Application) Thermal Desorption, Air Monitoring
Diameter (Metric) Outer 6.35 mm
Type Thermal Desorption Tube
Overall Length (Metric) 88.9 mm
Packaging Quantity 10/Pk.
For Use With (Equipment) PerkinElmer, Markes, DANI, OI Analytical, and Shimadzu Instruments
Overall Length (English) 3.5 in.
Diameter (English) Outer 1/4 in.
Includes Screens
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.