Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EMR2 Antibody (494014), CoraFluor™ 1, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB48941CL1
Description
EMR2 Monoclonal antibody specifically detects EMR2 in Human samples. It is validated for Flow CytometrySpecifications
| EMR2 | |
| Monoclonal | |
| CoraFluor 1 | |
| CD312, DKFZp781B135, egf-like module containing, mucin-like, hormone receptor-like 2, egf-like module containing, mucin-like, hormone receptor-like sequence 2, EGF-like module receptor 2, EGF-like module-containing mucin-like hormone receptor-like 2 | |
| NS0 mouse myeloma cell line transfected with human EMR2, Gln24-Asn823, Accession # AAI27006 | |
| 0.1 mL | |
| GPCR | |
| 30817 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b |
| Flow Cytometry | |
| 494014 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction