Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human EMR2 Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

$200.50 - $514.50

Specifications

Antigen EMR2
Clone 494014
Dilution Flow Cytometry 0.25 ug/106 cells
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB48941SP
GREENER_CHOICE
View Documents
R&D Systems
MAB48941SP
25 μg
Each for $200.50
Only null left
Add to Cart
 
MAB48941100
GREENER_CHOICE
View Documents
R&D Systems
MAB48941100
100 μg
Each for $514.50
Only null left
Add to Cart
 
Description

Description

EMR2 Monoclonal antibody specifically detects EMR2 in Human samples. It is validated for Flow Cytometry
Specifications

Specifications

EMR2
Flow Cytometry 0.25 ug/106 cells
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
CD312, DKFZp781B135, egf-like module containing, mucin-like, hormone receptor-like 2, egf-like module containing, mucin-like, hormone receptor-like sequence 2, EGF-like module receptor 2, EGF-like module-containing mucin-like hormone receptor-like 2
NS0 mouse myeloma cell line transfected with human EMR2, Gln24-Asn823, Accession # AAI27006
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
494014
Flow Cytometry
Unconjugated
Mouse
Human
AAI27006
30817
IgG2b
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.