Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EMR2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533505
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533505 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533505 Supplier Invitrogen™ Supplier No. PA533505
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human (100%) Gorilla (90%).

EMR3 gene encodes a member of the class B seven-span transmembrane (TM7) receptor family expressed predominantly by cells of the immune system. Family members are characterized by an extended extracellular region with a variable number of N-terminal epidermal growth factor (EGF)-like domains coupled to a TM7 domain via a mucin-like spacer domain. This gene is closely linked to the gene encoding egf-like molecule containing mucin-like hormone receptor 2 on chromosome 19. This protein may play a role in myeloid-myeloid interactions during immune and inflammatory responses.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EMR2
Applications ELISA, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene ADGRE2
Gene Accession No. Q9UHX3
Gene Alias ADGRE2; Adhesion G protein-coupled receptor E2; CD312; CD312 antigen; egf-like module containing, mucin-like, hormone receptor-like 2; EGF-like module receptor 2; EGF-like module-containing mucin-like hormone receptor-like 2; EMR2; VBU
Gene Symbols ADGRE2
Host Species Rabbit
Immunogen Synthetic 20 amino acid peptide from N-terminal extracellular domain of human EMR2.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30817
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.