Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EMR3 Antibody (3D7), PE/Cy5.5, Novus Biologicals™

Armenian Hamster Monoclonal Antibody
Supplier: Novus Biologicals NB10065935PECY55
Description
EMR3 Monoclonal specifically detects EMR3 in Human samples. It is validated for Flow Cytometry.Specifications
| EMR3 | |
| Monoclonal | |
| PE-Cyanine5.5 | |
| PBS with 0.05% Sodium Azide | |
| ADGRE3 | |
| ARHO-EMR3-CD97 (EGF1) transfectants. | |
| 0.1 mL | |
| Primary | |
| NB100-65935 recognizes human epidermal growth factor (EGF) module-containing mucin-like hormone receptor 3 (EMR3), which is a 56kD member of the EGF-7 transmembrane (TM7) family of adhesion receptors. EMR3 is expressed at the cell surface as a heterodimer. The molecule is predominantly expressed on granulocytes, and at lower levels on mature myeloid cells, monocytes and dendritic cells. EMR3 is absent on lymphocytes, haematopoietic stem cells and myeloid progenitors. Studies suggest that the EMR3 molecule is up-regulated during late stages of neutrophil differentiation and is a marker for terminally differentiated cells. The exact functions of EMR3 and its ligands have not yet been determined. | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| 3D7 | |
| Flow Cytometry | |
| egf-like module containing, mucin-like, hormone receptor-like 3, EGF-like module receptor 3, EGF-like module-containing mucin-like hormone receptor-like 3, egf-like module-containing mucin-like receptor 3 | |
| Armenian Hamster | |
| Protein G purified | |
| GPCR, Immunology, Innate Immunity | |
| 84658.0 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction