Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EMR3 Antibody (908235), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB8496MFV500
Description
EMR3 Monoclonal antibody specifically detects EMR3 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOFSpecifications
| EMR3 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| GPCR, Immunology, Innate Immunity | |
| 84658 | |
| Store at 4C in the dark. |
| Flow Cytometry, Immunohistochemistry, CyTOF | |
| 908235 | |
| Flow Cytometry, Immunohistochemistry, CyTOF-ready | |
| egf-like module containing, mucin-like, hormone receptor-like 3, EGF-like module receptor 3, EGF-like module-containing mucin-like hormone receptor-like 3, egf-like module-containing mucin-like receptor 3 | |
| NS0 mouse myeloma cell line transfected with human EMR3, Met1-Tyr652, Accession # Q9BY15 | |
| RUO | |
| Primary | |
| Human | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction