Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ EMR3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA533509
Description
Percent identity with other species by BLAST analysis: Human, Orangutan (100%) Marmoset, Elephant, Dog (80%).
EMR3 gene encodes a member of the class B seven-span transmembrane (TM7) receptor family expressed predominantly by cells of the immune system. Family members are characterized by an extended extracellular region with a variable number of N-terminal epidermal growth factor (EGF)-like domains coupled to a TM7 domain via a mucin-like spacer domain. This gene is closely linked to the gene encoding egf-like molecule containing mucin-like hormone receptor 2 on chromosome 19. This protein may play a role in myeloid-myeloid interactions during immune and inflammatory responses.
Specifications
EMR3 | |
Polyclonal | |
Unconjugated | |
ADGRE3 | |
ADGRE3; Adhesion G protein-coupled receptor E3; egf-like module containing, mucin-like, hormone receptor-like 3; EGF-like module receptor 3; EGF-like module-containing mucin-like hormone receptor-like 3; egf-like module-containing mucin-like receptor 3; EMR3; UNQ683/PRO1562 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84658 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q9BY15 | |
ADGRE3 | |
Synthetic 15 amino acid peptide from N-terminal extracellular domain of human EMR3. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction