Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EMR4 Polyclonal Antibody

EMR4 Polyclonal Antibody
Supplier: Thermo Scientific PA536374
Description
EMR4 Polyclonal Antibody for Western Blot, ICC/IF, IHC

Specifications
EMR4 | |
Polyclonal | |
Unconjugated | |
ADGRE4P | |
ADGRE4P; adhesion G protein-coupled receptor E4, pseudogene; EMR4; EMR4P; FIRE; GPR127; PGR16 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
326342 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.05% sodium azide; pH 7.2 | |
Q86SQ3 | |
ADGRE4P | |
Synthetic peptide corresponding to amino acids 108-164 of human EMR4. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction