Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EMSY Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP303288100UL
Description
EMSY Polyclonal specifically detects EMSY in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
EMSY | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunofluorescence | |
Unconjugated | |
chromosome 11 open reading frame 30, EMSYFLJ90741, protein EMSY | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1153-1322 of human EMSY (NP_064578.2). | |
100 μL | |
Apoptosis, Cancer, DNA Repair, Tumor Suppressors | |
56946 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction