Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Emx1 Antibody, Alexa Fluor™ 488, Novus Biologicals™

Chicken Polyclonal Antibody
Supplier: Novus Biologicals NBP241151AF488
Description
Emx1 Polyclonal antibody specifically detects Emx1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISASpecifications
Emx1 | |
Polyclonal | |
50mM Sodium Borate | |
Chicken | |
Peptide affinity purified | |
RUO | |
2016 | |
Store at 4°C in the dark. | |
IgY |
Western Blot, ELISA | |
Alexa Fluor 488 | |
empty spiracles homeobox 1, empty spiracles-like protein 1 | |
Antibody was raised against a 16 amino acid synthetic peptide near the amino terminus of human EMX1. The immunogen is located within amino acids 50 - 100 of EMX1. Amino Acid Squence: CAAAASEEPLRPTALN | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction