Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Emx1 Antibody - BSA Free, Novus Biologicals™

Chicken Polyclonal Antibody
Supplier: Novus Biologicals NBP2411510.025MG
Description
Emx1 Polyclonal specifically detects Emx1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.Specifications
Emx1 | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
EMX1 | |
Antibody was raised against a 16 amino acid synthetic peptide near the amino terminus of human EMX1. The immunogen is located within amino acids 50 - 100 of EMX1. Amino Acid Squence: CAAAASEEPLRPTALN | |
0.025 mg | |
Primary | |
At least two isoforms of EMX1 are known to exist; this antibody will detect the longer isoform. EMX1 antibody is predicted to not cross-react with other EMX family members. | |
Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
Western Blot, ELISA | |
1 mg/mL | |
Western Blot 1 μg/mL, ELISA | |
empty spiracles homeobox 1, empty spiracles-like protein 1 | |
Chicken | |
Affinity Purified | |
RUO | |
2016 | |
Human, Mouse, Rat | |
IgY |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction