Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Emx1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP313392
This item is not returnable.
View return policy
Description
Emx1 Polyclonal antibody specifically detects Emx1 in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
Emx1 | |
Polyclonal | |
PBS, 20% Glycerol (pH7) | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
2016 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
empty spiracles homeobox 1, empty spiracles-like protein 1 | |
Recombinant protein encompassing a sequence within the center region of mouse Emx1. The exact sequence is proprietary. | |
100 μg | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction