Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    Emx1 Antibody, PerCP, Novus Biologicals™
 
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Chicken Polyclonal Antibody
Supplier: Novus Biologicals NBP241151PCP
Description
Emx1 Polyclonal antibody specifically detects Emx1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISASpecifications
| Emx1 | |
| Polyclonal | |
| Western Blot, ELISA | |
| empty spiracles homeobox 1, empty spiracles-like protein 1 | |
| Antibody was raised against a 16 amino acid synthetic peptide near the amino terminus of human EMX1. The immunogen is located within amino acids 50 - 100 of EMX1. Amino Acid Squence: CAAAASEEPLRPTALN | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified | 
| Western Blot, ELISA | |
| PerCP | |
| PBS | |
| Chicken | |
| Peptide affinity purified | |
| RUO | |
| 2016 | |
| Store at 4C in the dark. | |
| IgY | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
            